1,6-Dibromohexane ≥95%

Supplier: Matrix Scientific

075625-10G 075625-100G
102542-214EA 36.92 USD
102542-214 102542-216
1,6-Dibromohexane ≥95%
1,6-Dibromohexane

Hazard: Toxic

Formula: BrH₂C(CH₂)₄CH₂Br
Boiling Pt: 243 °C
Density: 1.586
Flash Pt: 113 °C
MDL Number: MFCD00000272
CAS Number: 629-03-8
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR