2-Chloro-(4-methoxycarbonyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

075621-1G 075621-5G 075621-10G
102542-194EA 115.38 USD
102542-194 102542-196 102542-198
2-Chloro-(4-methoxycarbonyl)phenylboronic acid ≥95%
2-Chloro-(4-methoxycarbonyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₈BClO₄
MDL Number: MFCD08689530
CAS Number: 603122-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR