3-Oxoglutaric acid ≥95%

Supplier: Matrix Scientific

075619-10G 075619-100G
102542-186EA 36.92 USD
102542-186 102542-188
3-Oxoglutaric acid ≥95%
3-Oxoglutaric acid

Hazard: Irritant

Formula: C₅H₆O₅
Melting Pt: 136-9°C (133°C)
MDL Number: MFCD00002711
CAS Number: 542-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR