1,9-Dibromononane ≥95%

Supplier: Matrix Scientific

Synonyms: Nonamethylene dibromide

075618-10G 075618-100G
102542-182EA 641.54 USD
102542-182 102542-184
1,9-Dibromononane ≥95%
1,9-Dibromononane

Hazard: Irritant

Formula: BrH₂C(CH₂)₇CH₂Br
MDL Number: MFCD00000279
CAS Number: 4549-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR