4-Methyl-1-naphthoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-1-naphthalenecarboxylic acid

075617-1G 075617-5G 075617-10G
102542-176EA 72.31 USD
102542-176 102542-178 102542-180
4-Methyl-1-naphthoic acid ≥95%
4-Methyl-1-naphthoic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00671557
CAS Number: 4488-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR