(E)-Ethyl-2-cyano-3-ethoxyacrylate ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-Ethyl 2-cyano-3-ethoxyacrylate

075616-1G 075616-5G 075616-10G
102542-170EA 107.69 USD
102542-170 102542-172 102542-174
(E)-Ethyl-2-cyano-3-ethoxyacrylate ≥95%
(E)-Ethyl-2-cyano-3-ethoxyacrylate

Hazard: Irritant

Formula: C₈H₁₁NO₃
MW: 169.1778 g/mol
MDL Number: MFCD00009136
CAS Number: 42466-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR