Toluene-p-sulfonic anhydride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride

075615-100G 075615-25G
102542-168EA 227.69 USD
102542-168 102833-858
Toluene-p-sulfonic anhydride ≥95%
Toluene-p-sulphonic anhydride

Hazard: Irritant

Formula: C₁₄H₁₄O₅S₂
MW: 326.39 g/mol
Melting Pt: 121-130
MDL Number: MFCD00008548
CAS Number: 4124-41-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR