(R)-2-Amino-2-phenylacetyl chloride hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: D-(-)-2-Phenylglycine chloride hydrochloride

075613-1G 075613-5G 075613-10G
102542-154EA 43.08 USD
102542-154 102542-156 102542-158
(R)-2-Amino-2-phenylacetyl chloride hydrochloride ≥95%
(R)-2-Amino-2-phenylacetyl chloride hydrochloride

Hazard: Irritant

Formula: C₈H₉Cl₂NO
MDL Number: MFCD00039094
CAS Number: 39878-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR