Ethyl-4-chlorobenzoylformate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(4-chlorophenyl)-2-oxoacetate, Ethyl (4-chlorophenyl)(oxo)acetate
075609-1G
075609-5G
075609-10G
102542-132EA
61.54
USD
102542-132
102542-134
102542-136
Ethyl-4-chlorobenzoylformate ≥95%
Ethyl 4-chlorobenzoylformate
Hazard: Irritant
Formula:
C₁₀H₉ClO₃ |
MDL Number:
MFCD01319615 CAS Number: 34966-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...