4-Iodo-L-phenylalanine ≥95%
Supplier: Matrix Scientific
Synonyms:
H-Phe(4-I)-OH
075606-5G
075606-25G
075606-100G
102542-120EA
73.85
USD
102542-120
102833-854
102833-856
4-Iodo-L-phenylalanine ≥95%
4-Iodo-L-phenylalanine
Hazard: Irritant-Harmful/Store in Freezer/Light Sensitive
Formula:
C₉H₁₀INO₂ MW: 291.1 g/mol |
MDL Number:
MFCD00002602 CAS Number: 24250-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...