2'-Bromoacetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
o-Bromoacetophenone
075605-10G
075605-100G
102542-114EA
72.31
USD
102542-114
102542-116
2'-Bromoacetophenone ≥95%
2'-Bromoacetophenone
Hazard: Irritant
Formula:
C₈H₇BrO MW: 199.05 g/mol Density: 1.476 |
MDL Number:
MFCD00000067 CAS Number: 2142-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...