4-Chloro-6-methoxyquinolin-7-ol ≥95%

Supplier: Matrix Scientific

075604-1G 075604-5G
102542-110EA 558.46 USD
102542-110 102542-112
4-Chloro-6-methoxyquinolin-7-ol ≥95%
4-Chloro-6-methoxyquinolin-7-ol

Hazard: Irritant

Formula: C₁₀H₈ClNO₂
MW: 209.6290 g/mol
MDL Number: MFCD13191788
CAS Number: 205448-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR