2-Cyanopyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile

075601-10G 075601-100G
102542-096EA 36.92 USD
102542-096 102542-098
2-Cyanopyrazine ≥95%
2-Cyanopyrazine

Hazard: Irritant

Formula: C₅H₃N₃
MW: 105.10 g/mol
MDL Number: MFCD00049361
CAS Number: 19847-12-2
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR