(4-Chlorophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chlorobenzyl cyanide

075597-10G 075597-100G
102542-080EA 300 USD
102542-080 102542-082
(4-Chlorophenyl)acetonitrile ≥95%
(4-Chlorophenyl)acetonitrile

Hazard: Irritant

Formula: C₈H₆ClN
MW: 151.6 g/mol
MDL Number: MFCD00001918
CAS Number: 140-53-4
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR