1-Methyl-1H-imidazole-4-carboxamide ≥95%

Supplier: Matrix Scientific

075596-250MG 075596-1G 075596-5G
102542-074EA 355.38 USD
102542-074 102542-076 102542-078
1-Methyl-1H-imidazole-4-carboxamide ≥95%
1-Methyl-1H-imidazole-4-carboxamide

Hazard: Irritant

Formula: C₅H₇N₃O
CAS Number: 129993-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR