7-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine ≥95%

Supplier: Matrix Scientific

075592-1G 075592-5G
102542-062EA 569.23 USD
102542-062 102542-064
7-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine ≥95%
7-Chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine

Hazard: Irritant

Formula: C₈H₈ClNO
MW: 169.6082 g/mol
MDL Number: MFCD07364075
CAS Number: 113770-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR