2-Bromo-3-nitropyridine-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-bromo-3-nitropyridine

075591-250MG 075591-1G 075591-5G
102542-056EA 323.08 USD
102542-056 102542-058 102542-060
2-Bromo-3-nitropyridine-4-amine ≥95%
2-Bromo-3-nitropyridine-4-amine

Hazard: Irritant

Formula: C₅H₄BrN₃O₂
MW: 218.0100 g/mol
MDL Number: MFCD11656226
CAS Number: 84487-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR