Methyl-3,5-dimethoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3,5-dimethoxybenzoate, 3,5-Dimethoxybenzoic acid methyl ester

075590-10G 075590-100G
102542-052EA 64.62 USD
102542-052 102542-054
Methyl-3,5-dimethoxybenzoate ≥95%
Methyl-3,5-dimethoxybenzoate

Hazard: Irritant

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
MDL Number: MFCD00008432
CAS Number: 2150-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR