Methyl-3-fluorobenzoate ≥95%

Supplier: Matrix Scientific

075589-10G 075589-100G
102542-048EA 30.77 USD
102542-048 102542-050
Methyl-3-fluorobenzoate ≥95%
Methyl-3-fluorobenzoate

Hazard: Irritant

Formula: C₈H₇FO₂
MDL Number: MFCD03094302
CAS Number: 455-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR