Lapatinib ditosylate ≥95%

Supplier: Matrix Scientific

Synonyms: Tyverb, N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulphonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulphonate), Tykerb, N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulfonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulphonate), GW572016 ditosylate, GW2016 ditosylate

075587-1G 075587-5G
102542-038EA 961.54 USD
102542-038 102542-040
Lapatinib ditosylate ≥95%
Lapatinib ditosylate

Hazard: Irritant

Formula: C₂₉H₂₆ClFN₄O₄S·₂C₇H₈O₃S
MW: 925.46 g/mol
MDL Number: MFCD09264195
CAS Number: 388082-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR