4-(Tetrahydropyran-4-yl)phenylamine ≥95%

Supplier: Matrix Scientific

075586-250MG 075586-1G 075586-5G
102542-032EA 598.46 USD
102542-032 102542-034 102542-036
4-(Tetrahydropyran-4-yl)phenylamine ≥95%
4-(Tetrahydropyran-4-yl)phenylamine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD11506022
CAS Number: 62071-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR