2,5-Dichlorobenzoic acid ≥95%

Supplier: Matrix Scientific

075585-10G 075585-100G
102542-028EA 72.31 USD
102542-028 102542-030
2,5-Dichlorobenzoic acid ≥95%
2,5-Dichlorobenzoic acid

Hazard: Irritant

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
MDL Number: MFCD00002416
CAS Number: 50-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR