Fmoc-Thr-OH ≥95%

Supplier: Matrix Scientific

Synonyms: (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid, Fmoc-L-Threonine

075584-10G 075584-100G
102542-024EA 38.46 USD
102542-024 102542-026
Fmoc-Thr-OH ≥95%
Fmoc-Thr-OH

Hazard: Irritant

Formula: C₁₉H₁₉NO₅
MW: 341.358 g/mol
MDL Number: MFCD00077072
CAS Number: 73731-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR