3-Methoxysalicylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-3-methoxybenzoic acid
075583-10G
075583-100G
102542-020EA
64.62
USD
102542-020
102542-022
3-Methoxysalicylic acid ≥95%
3-Methoxysalicylic acid
Hazard: Irritant
Formula:
C₈H₈O₄ MW: 168.15 g/mol |
MDL Number:
MFCD00002445 CAS Number: 877-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...