3,4-Methylenedioxyphenethylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[3,4-(Methylenedioxy)phenyl]ethylamine

075580-100G 075580-25G
102542-006EA 692.31 USD
102542-006 102833-852
3,4-Methylenedioxyphenethylamine ≥95%
3,4-Methylenedioxyphenethylamine

Hazard: Irritant

Formula: C9H11NO2
MW: 165.19 g/mol
MDL Number: MFCD00060509
CAS Number: 1484-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR