(4-Pyridyl)acetone ≥95%

Supplier: Matrix Scientific

075579-1G 075579-5G 075579-10G
102541-998EA 15.38 USD
102541-998 102542-000 102542-002
(4-Pyridyl)acetone ≥95%
(4-Pyridyl)acetone

Hazard: Irritant

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 143°C/20mm
Melting Pt: 13 °C
MDL Number: MFCD00129043
CAS Number: 6304-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR