5-Chloro-2-methoxybenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-o-anisic acid

075578-10G 075578-100G 075578-500G
102541-992EA 38.46 USD
102541-992 102541-994 102541-996
5-Chloro-2-methoxybenzoic acid ≥95%
5-Chloro-2-methoxybenzoic acid

Hazard: Irritant

Formula: C₈H₇ClO₃
MDL Number: MFCD00017546
CAS Number: 3438-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR