Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific
Warning

075574-5G 075574-10G 075574-100G
102541-972EA 107.69 USD
102541-972 102541-974 102541-976
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride ≥95%
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

Purity: 95+% CAS: 1454-53-1 MDL Number: MFCD00012799 Molecular Formula: C14H18ClNO3 Molecular Weight: 283,76 Hazard: Irritant

Formula: C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl
MW: 283.75 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00012799
CAS Number: 3939-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR