3,5-Dimethoxybenzyl alcohol ≥95%

Supplier: Matrix Scientific

Synonyms: (3,5-Dimethoxyphenyl)methanol

075573-10G 075573-100G
102541-968EA 107.69 USD
102541-968 102541-970
3,5-Dimethoxybenzyl alcohol ≥95%
3,5-Dimethoxybenzyl alcohol

Hazard: Irritant

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00004641
CAS Number: 705-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR