4-Chloro-6-ethyl-5-fluoropyrimidine ≥95%

Supplier: Matrix Scientific

075571-5G 075571-100G
102541-956EA 35.38 USD
102541-956 102541-960
4-Chloro-6-ethyl-5-fluoropyrimidine ≥95%
4-Chloro-6-ethyl-5-fluoropyrimidine

Hazard: Irritant

Formula: C₆H₆ClFN₂
MDL Number: MFCD07782087
CAS Number: 137234-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR