5,6,7,8-Tetrahydro-2-naphthol ≥95%

Supplier: Matrix Scientific

075570-10G 075570-100G
102541-952EA 27.69 USD
102541-952 102541-954
5,6,7,8-Tetrahydro-2-naphthol ≥95%
5,6,7,8-Tetrahydro-2-naphthol

Hazard: Irritant

Formula: C₁₀H₁₂O
Boiling Pt: 275…276 °C
Melting Pt: 60…63 °C
Flash Pt: >110 °C
MDL Number: MFCD00001738
CAS Number: 1125-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR