cis-4-Cyclohexene-1,2-dicarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (1R,2S)-rel-Cyclohex-4-ene-1,2-dicarboxylic acid

075569-10G 075569-100G
102541-948EA 47.69 USD
102541-948 102541-950
cis-4-Cyclohexene-1,2-dicarboxylic acid ≥95%
cis-4-Cyclohexene-1,2-dicarboxylic acid

Hazard: Irritant

Formula: C₈H₁₀O₄
MW: 170.17 g/mol
MDL Number: MFCD00070482
CAS Number: 2305-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR