1,6-Dihydroxynaphthalene ≥95%

Supplier: Matrix Scientific

Synonyms: Naphthalene-1,6-diol

075568-10G 075568-100G
102541-944EA 38.46 USD
102541-944 102541-946
1,6-Dihydroxynaphthalene ≥95%
1,6-Dihydroxynaphthalene

Hazard: Irritant

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD00003981
CAS Number: 575-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR