(R)-(+)-β-Methylphenethylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-(+)-beta-Methylphenethylamine, (R)-(+)-2-Phenyl-1-propylamine
075565-1G
075565-5G
075565-10G
102541-934EA
61.54
USD
102541-934
102541-936
102541-938
(R)-(+)-β-Methylphenethylamine ≥95%
(R)-(+)-β-Methylphenethylamine
Hazard: Corrosive/Irritant-Harmful/Stored under Argon
Formula:
C₉H₁₃N Boiling Pt: 197°C Melting Pt: 81°C Density: 0.945 |
MDL Number:
MFCD00216740 CAS Number: 28163-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...