4-(N,N-Diphenylamino)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Diphenylaminobenzaldehyde

075564-1G 075564-5G 075564-10G
102541-928EA 72.31 USD
102541-928 102541-930 102541-932
4-(N,N-Diphenylamino)benzaldehyde ≥95%
4-(N,N-Diphenylamino)benzaldehyde

TCA listed: T
Hazard: Irritant

Formula: C₁₉H₁₅NO
MW: 273.33 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00145131
CAS Number: 4181-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR