Pimelic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Heptanedioic acid, 1,5-Pentanedicarboxylic acid, 1,7-Heptanedioic acid

075563-10G 075563-100G
102541-924EA 55.38 USD
102541-924 102541-926
Pimelic acid ≥95%
Pimelic acid

Hazard: Irritant

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
MDL Number: MFCD00004425
CAS Number: 111-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR