2-Piperidinemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)piperidine

075562-10G 075562-100G
102541-920EA 72.31 USD
102541-920 102541-922
2-Piperidinemethanol ≥95%
2-Piperidinemethanol

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD00005987
CAS Number: 3433-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR