Boc-Glu-OH ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Glu-OH, N-Boc-L-glutamic Acid

075561-10G 075561-100G
102541-916EA 90.77 USD
102541-916 102541-918
Boc-Glu-OH ≥95%
Boc-Glu-OH

Hazard: Irritant

Formula: C₁₀H₁₇NO₆
MDL Number: MFCD00037297
CAS Number: 2419-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR