2-Bromo-3-chloro-5-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-5-chloropyridin-3-ol

075558-250MG 075558-1G 075558-5G
102541-904EA 684.62 USD
102541-904 102541-906 102541-908
2-Bromo-3-chloro-5-hydroxypyridine ≥95%
2-Bromo-3-chloro-5-hydroxypyridine

Hazard: Irritant

Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
MDL Number: MFCD09835244
CAS Number: 130284-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR