Methyl-3-benzamidopropanoate ≥95%

Supplier: Matrix Scientific

075550-1G 075550-5G 075550-10G
102541-862EA 178.46 USD
102541-862 102541-864 102541-866
Methyl-3-benzamidopropanoate ≥95%
Methyl-3-benzamidopropanoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
CAS Number: 89928-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR