Boc-L-tyrosine methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Tyr-OMe, N-Boc-L-tyrosine Methyl Ester

075548-100G 075548-25G
102541-856EA 104.62 USD
102541-856 102551-976
Boc-L-tyrosine methyl ester ≥95%
Boc-L-tyrosine methyl ester

Hazard: Irritant

Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Melting Pt: 105…108 °C
MDL Number: MFCD00191181
CAS Number: 4326-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR