trans-Methyl-4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: trans-Methyl 4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate

075547-1G 075547-5G
102541-848EA 90.77 USD
102541-848 102541-850
trans-Methyl-4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate ≥95%
trans-Methyl-4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD17015041
CAS Number: 146307-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR