4-Iodo-2-nitrotoluene ≥95%

Supplier: Matrix Scientific

075538-5G 075538-25G
102541-812EA 55.38 USD
102541-812 102833-848
4-Iodo-2-nitrotoluene ≥95%
4-Iodo-2-nitrotoluene

Hazard: Irritant

Formula: C₇H₆INO₂
Melting Pt: 58…62 °C
MDL Number: MFCD00051090
CAS Number: 41252-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR