Trimethyl ethane-1,1,2-tricarboxylate ≥95%

Supplier: Matrix Scientific

075537-1G 075537-5G 075537-10G
102541-806EA 30.77 USD
102541-806 102541-808 102541-810
Trimethyl ethane-1,1,2-tricarboxylate ≥95%
Trimethyl ethane-1,1,2-tricarboxylate

Hazard: Irritant

Formula: C₈H₁₂O₆
MW: 204.1800 g/mol
MDL Number: MFCD00015619
CAS Number: 40967-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR