(4-Methoxyphenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxyphenylacetic acid

075531-10G 075531-100G
102541-778EA 36.92 USD
102541-778 102541-780
(4-Methoxyphenyl)acetic acid ≥95%
(4-Methoxyphenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
MDL Number: MFCD00004345
CAS Number: 104-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR