3,4-Dimethylphenylhydrazinium chloride ≥95%

Supplier: Matrix Scientific

Synonyms: (3,4-Dimethylphenyl)hydrazine hydrochloride, 3,4-Dimethylphenylhydrazine hydrochloride, 1-(3,4-Dimethylphenyl)hydrazine hydrochloride

075530-10G 075530-100G
102541-774EA 107.69 USD
102541-774 102541-776
3,4-Dimethylphenylhydrazinium chloride ≥95%
3,4-Dimethylphenylhydrazinium chloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
MDL Number: MFCD00052270
CAS Number: 60481-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR