4'-Pentylacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: p-Pentylacetophenone, 4-Pentylacetophenone

075529-10G 075529-100G
102541-770EA 178.46 USD
102541-770 102541-772
4'-Pentylacetophenone ≥95%
4'-Pentylacetophenone

Hazard: Irritant

Formula: C₁₃H₁₈O
MW: 190.29
MDL Number: MFCD00043687
CAS Number: 37593-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR