3-Nitrophthalic anhydride ≥95%

Supplier: Matrix Scientific

075528-10G 075528-100G
102541-766EA 13.08 USD
102541-766 102541-768
3-Nitrophthalic anhydride ≥95%
3-Nitrophthalic anhydride

Hazard: Irritant

Formula: C₈H₃NO₅
Melting Pt: 164 °C
Density: 1.364
MDL Number: MFCD00005921
CAS Number: 641-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR