Methyl-3-cyclopentenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-cyclopentenecarboxylate

075526-5G 075526-25G 075526-100G
102541-760EA 52.31 USD
102541-760 102551-972 102551-974
Methyl-3-cyclopentenecarboxylate ≥95%
Methyl-3-cyclopentenecarboxylate

Hazard: Flammable

Formula: C₇H₁₀O₂
MW: 126.15 g/mol
Density: 1.031
Flash Pt: 48.9 °C
MDL Number: MFCD04038661
CAS Number: 58101-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR