L-(-)-tert-Leucine (H-Tle-OH) ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH
075521-5G
075521-25G
102541-736EA
21.54
USD
102541-736
102833-846
L-(-)-tert-Leucine (H-Tle-OH) ≥95%
L(-)-tert-Leucine (H-Tle-OH)
Hazard: Irritant
Formula:
C₆H₁₃NO₂ MW: 131.2 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00064218 CAS Number: 20859-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...